Test sieves, 200×25 mm
Leverancier: ENDECOTTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
200SIW.100H 200SIW.200H 200SIW.300H 200SIW.400H 200SIW.500H 200SIW.600H 200SIW.710H 200SIW.800H 200SIW.900H 200SIW1.00H 200SIW1.18H 200SIW.212H 200SIW.150H 200SIW.180H 200SIW.250H 200SIW.063H 200SIW.425H 200SIW.850H 200SIW1.40H 200SIW1.60H 200SIW4.00H 200SIW2.80H 200SIW5.60H 200SIW8.00H 200SIW.355H 200SIW.125H 200SIW2.00H 200SIW.045H 200SIW.090H 200SIW.106H 200SIW.050CH 200SIW.150C 200SIW.106C 200SIW.425C 200SIW.600CH 200SIW1.00CH 200SIW10.0CH
ENDE200SIW.100HEA 209 EUR
ENDE200SIW.100H ENDE200SIW.200H ENDE200SIW.300H ENDE200SIW.400H ENDE200SIW.500H ENDE200SIW.600H ENDE200SIW.710H ENDE200SIW.800H ENDE200SIW.900H ENDE200SIW1.00H ENDE200SIW1.18H ENDE200SIW.212H ENDE200SIW.150H ENDE200SIW.180H ENDE200SIW.250H ENDE200SIW.063H ENDE200SIW.425H ENDE200SIW.850H ENDE200SIW1.40H ENDE200SIW1.60H ENDE200SIW4.00H ENDE200SIW2.80H ENDE200SIW5.60H ENDE200SIW8.00H ENDE200SIW.355H ENDE200SIW.125H ENDE200SIW2.00H ENDE200SIW.045H ENDE200SIW.090H ENDE200SIW.106H ENDE200SIW.050CH ENDE200SIW.150C ENDE200SIW.106C ENDE200SIW.425C ENDE200SIW.600CH ENDE200SIW1.00CH ENDE200SIW10.0CH
Test sieves, 200×25 mm
Zeven Testzeven
Stainless steel woven wire mesh test sieves are ideal for all types of laboratory sampling and particle size analysis.
For more size options please contact your local Avantor office.
* Calibrated test sieves: Test sieves inspected and calibrated in accordance with procedures listed in clause 5.2 and table 4 of ISO 3310-1 and BS 410-1. Each sieve is supplied with a calibration certificate recording the number of aperture and wire diameters measured, the average aperture size and standard deviation separately for the warp and weft direction. The type of weave will also be stated.