Spray nozzles
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
215-66 215-15 215-1 215-28 215-14
ENTE215-66EA 0 EUR
ENTE215-66 ENTE215-15 ENTE215-1 ENTE215-28 ENTE215-14
Spray nozzles
Spuitpistolen
PFA spray nozzles with NPT connections for use in applications needing chemical or DI water to be sprayed across a surface.
Molded PFA provides universal chemical compatibility and use in ultrapure and chemically corrosive applications.