Tank, rectangular, PFA
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E92-01D-0215
ENTEE92-01D-0215EA 0 EUR
ENTEE92-01D-0215
Tank, rectangular, PFA
Reservoirs Rechthoekige tanks
Injection molded rectangular tanks with dual capacity.
  • Available in single or dual capacity
  • Available with or without covers
  • Tanks should be properly supported in heated applications
Maximum recommended working temperature: 180 °C.
Mounting hole dimension (L×W): 419×216 mm.