Trays, H 20 Series
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
H204502562C02 H201902462C02 H2013018262C02 H200902466C02 H2010411966C02 H2011014562C02 H20-430-62C02U H2006365066C02 H200452466C02 H2007410462C02 H200752462C02 H200952462C02 H2010014562C02 H2010916866C02 H201202466C02 H201502462C02 H2015950962C02 H2016921962C02 H20-219-62C02 H20-300-1415 H20-411-62C02 H20-551-66C02 H20-300-62C02 H20-300-66C02 H200501666C02 H20-315-62C02 H2009013062C02 H200301662C02 H20-229-62C02 H202502462C02 H20-551-1415 H2004508062C02 H2005206266C02 H200641966C02 H2007410466C02 H20095-24-1415 H2010014566C02 H2025628862C02 H2040030062C02 H20-411-1415 H2031046066C02 H2008509862C02 H2052211066C02 H20N33070866C2 H20-169-62C02 H201402462C02 H2018127162C02 H200805062C02 H20-450-62C02U H2034544562C02 H20N72250862C2 H202202462C02 H2004105766C02 H20E17528566C2 H2057328466C02 H2018144562C02 H202855062C02 H20-485-66C02 H2010012062C02 H2025123062C02 H2023087066C02 H2003206766C02 H200302462C02 H200757562C02 H2002806066C02 H20-811-66C02 H20-600WH-62C2 H2002304566C02 H2052100766C02 H20-165-66C02 H200955162C02 H201154562C02 H20E16010066C2 H2015933466C02 H2003705166C02 H2003705162C02 H2003001766C02 H20N21410266C2 H200371566C02 H204146066C02 H2025535766C02 H20-035-66C02 H201502466C02 H20-1252301415 H20-0600901415 H20-0321566C02 H20-2101666C02 H2031032-62C02 H2003806666C02 H2008008562C02 H2004107366C02 H2007517566C02 H2011013162C02 H2003817066C02 H2005409062C02 H200261262C02 H200261266C02 H200261562C02 H200261566C02 H200270471415 H2002704762C02 H20-028-66C02 H2002905366C02 H200302466C02 H200303066C02 H2003124266C02 H200340866C02 H2003503766C02 H2003504566C02 H2003505062C02 H2003505066C02 H20-035-62C02 H2003606866C02 H200361466C02 H200371562C02 H20-037-62C02 H20-037-66C02 H200502466C02 H2040030066C02 H2002904366C02 H204146062C02 H20E17215966C0 H2055024TW66C0 H2016012066C02 H200653566C02U H201244366C02 H2003006562C02 H20-077-66C02 H20E0873266C02 H20I10111366C2
ENTEH204502562C02EA 0 EUR
ENTEH204502562C02 ENTEH201902462C02 ENTEH2013018262C02 ENTEH200902466C02 ENTEH2010411966C02 ENTEH2011014562C02 ENTEH20-430-62C02U ENTEH2006365066C02 ENTEH200452466C02 ENTEH2007410462C02 ENTEH200752462C02 ENTEH200952462C02 ENTEH2010014562C02 ENTEH2010916866C02 ENTEH201202466C02 ENTEH201502462C02 ENTEH2015950962C02 ENTEH2016921962C02 ENTEH20-219-62C02 ENTEH20-300-1415 ENTEH20-411-62C02 ENTEH20-551-66C02 ENTEH20-300-62C02 ENTEH20-300-66C02 ENTEH200501666C02 ENTEH20-315-62C02 ENTEH2009013062C02 ENTEH200301662C02 ENTEH20-229-62C02 ENTEH202502462C02 ENTEH20-551-1415 ENTEH2004508062C02 ENTEH2005206266C02 ENTEH200641966C02 ENTEH2007410466C02 ENTEH20095-24-1415 ENTEH2010014566C02 ENTEH2025628862C02 ENTEH2040030062C02 ENTEH20-411-1415 ENTEH2031046066C02 ENTEH2008509862C02 ENTEH2052211066C02 ENTEH20N33070866C2 ENTEH20-169-62C02 ENTEH201402462C02 ENTEH2018127162C02 ENTEH200805062C02 ENTEH20-450-62C02U ENTEH2034544562C02 ENTEH20N72250862C2 ENTEH202202462C02 ENTEH2004105766C02 ENTEH20E17528566C2 ENTEH2057328466C02 ENTEH2018144562C02 ENTEH202855062C02 ENTEH20-485-66C02 ENTEH2010012062C02 ENTEH2025123062C02 ENTEH2023087066C02 ENTEH2003206766C02 ENTEH200302462C02 ENTEH200757562C02 ENTEH2002806066C02 ENTEH20-811-66C02 ENTEH20-600WH-62C2 ENTEH2002304566C02 ENTEH2052100766C02 ENTEH20-165-66C02 ENTEH200955162C02 ENTEH201154562C02 ENTEH20E16010066C2 ENTEH2015933466C02 ENTEH2003705166C02 ENTEH2003705162C02 ENTEH2003001766C02 ENTEH20N21410266C2 ENTEH200371566C02 ENTEH204146066C02 ENTEH2025535766C02 ENTEH20-035-66C02 ENTEH201502466C02 ENTEH20-1252301415 ENTEH20-0600901415 ENTEH20-0321566C02 ENTEH20-2101666C02 ENTEH2031032-62C02 ENTEH2003806666C02 ENTEH2008008562C02 ENTEH2004107366C02 ENTEH2007517566C02 ENTEH2011013162C02 ENTEH2003817066C02 ENTEH2005409062C02 ENTEH200261262C02 ENTEH200261266C02 ENTEH200261562C02 ENTEH200261566C02 ENTEH200270471415 ENTEH2002704762C02 ENTEH20-028-66C02 ENTEH2002905366C02 ENTEH200302466C02 ENTEH200303066C02 ENTEH2003124266C02 ENTEH200340866C02 ENTEH2003503766C02 ENTEH2003504566C02 ENTEH2003505062C02 ENTEH2003505066C02 ENTEH20-035-62C02 ENTEH2003606866C02 ENTEH200361466C02 ENTEH200371562C02 ENTEH20-037-62C02 ENTEH20-037-66C02 ENTEH200502466C02 ENTEH2040030066C02 ENTEH2002904366C02 ENTEH204146062C02 ENTEH20E17215966C0 ENTEH2055024TW66C0 ENTEH2016012066C02 ENTEH200653566C02U ENTEH201244366C02 ENTEH2003006562C02 ENTEH20-077-66C02 ENTEH20E0873266C02 ENTEH20I10111366C2
Trays, H 20 Series
Wafertransportsystemen Wafer dragers
Entegris offers a wide selection of H 20 series 2" square trays, covers and accessories to meet your changing needs. H 20 series trays are available in various pocket sizes and depths, with or without wash holes and tweezers slots. H 20 series products offer proven reliability for handling bare die, including:
  • Known good die (KGD)
  • Wafer scale packing (WSP)
  • Chip scal packing (CSP)
  • Flip chip
  • Bump die
  • Opto electronics
  • Passive and active chip components, mechanical components