Testosterone high sensitivity ELISA kit

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ADI900176 ADI901176
ENZOADI900176EA 495 EUR
ENZOADI900176 ENZOADI901176
Testosterone high sensitivity ELISA kit
Testen ELISAs

Testosterone ELISA kit is a highly sensitive competitive immunoassay for the measurement of testosterone in human serum and plasma samples. Results are read on a colorimetric plate reader at 405 nm.


  • Excellent sensitivity
  • Validated for use with human samples
  • Stable, colour coded reagents reduce errors


Leveringsinformatie: Kit includes GxM microtiter plate, assay buffer, standard, antibody, conjugate, wash buffer concentrate, pNpp substrate, stop solution.

New Product

Order Now

Specificaties

Environmentally Preferable
Target protein Testosterone
Primary antibody reactivity Human
Assaytype Competitive
Host
Gevoeligheid 2,6 pg/ml (range 3,9 - 1000 pg/ml)
Storage conditions 4 °C (short and long term)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR