Human recombinant HSF1 (from E. coli), HIS tag
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ADISPP900F
ENZOADISPP900FEA 732 EUR
ENZOADISPP900F
Human recombinant HSF1 (from E. coli), HIS tag
Eiwitten en peptiden
Produced in E. coli. Human HSF1 is fused at the N-terminus to a His-tag.