Anti-GRP75 Mouse Monoclonal Antibody

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ADISPS825D ADISPS825F
ENZOADISPS825DEA 422 EUR
ENZOADISPS825D ENZOADISPS825F
Anti-GRP75 Mouse Monoclonal Antibody
Antilichamen
Grp75 (Glucose regulated 75 kDa protein) is a member of the Hsp70 family. It is localized in the mitochondrial matrix, where, in concert with Hsp60, is thought to participate in the re-folding of proteins translocated into this organelle. Like its E. coli homolog DnaK, Grp75 possesses a cation-dependent ATPase activity thought to be central to its function as a chaperone.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, Drosophila, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus Grp75. Detects aMore Product Information
Order Now

Specificaties

antigeen symbool GRP75
Type antilichaam Primair,Glucose-regulated Protein 75kDa
Environmentally Preferable
Type antilichaam Primair,Glucose-regulated Protein 75kDa
Clonality Monoklonaal
Reactivity
Host Mouse
Isotype IgG1
Western blot Yes
ImmunoChemie Yes
Formulier Liquid. In PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide.
Purification Ammonium sulfate precipitate

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR