Human MMP8 proenzyme (from neutrophils)
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ALX200421C005
ENZOALX200421C005EA 146 EUR
ENZOALX200421C005
Human MMP8 proenzyme (from neutrophils)
Eiwitten en peptiden
MMP-8 is a glycoprotein containing complex N-linked oligosaccharides.
  • Hydrolyzes type I over type II, and III collagens
  • Activated MMP-8 is inhibited by TIMP-1 (Prod. No. ALX-200-426)
Isolated from stimulated human neutrophil granulocytes (buffy coat). Requires activation.