Suc-Leu-Tyr-AMC
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ALX260054M005
ENZOALX260054M005EA 50.6 EUR
ENZOALX260054M005
Suc-Leu-Tyr-AMC
Eiwitten en peptiden
Protease substrate
Substrate for porcine calpain I and II. Also cleaved by papain and E. coli protease Ti. Ex.: 340 to 360 nm, Em.: 440 to 460 nm. This substrate is useful for inhibitor screening and kinetic analysis.