PD 184,352 ≥95% (by HPLC, NMR)

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ALX270471M001 ALX270471M005
ENZOALX270471M001EA 149 EUR
ENZOALX270471M001 ENZOALX270471M005
PD 184,352 ≥95% (by HPLC, NMR)
PD 184,352

Potent and selective inhibitor of MAPK (ERK kinase 1; MEK1) activation (IC50=300nM in vitro, IC50=2nM in vivo). Suppresses activation of MAPK but does not block its activity. Strongly suppresses growth of human implanted colon tumors in mice.


Synthetic.

Formule: C₁₇H₁₄ClF₂IN₂O₂
MDL Nummer: MFCD02683961
CAS nummer: 212631-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR