Gentamicin sulphate

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

ALX380003G001 ALX380003G005 ALX380003G010 ALX380003G025
ENZOALX380003G001EA 83.6 EUR
ENZOALX380003G001 ENZOALX380003G005 ENZOALX380003G010 ENZOALX380003G025
Gentamicin sulphate
Gentamicin sulphate
Smeltpunt: 218…237 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00270181
CAS nummer: 1405-41-0
EINECS: 215-778-9
Merck Index: 13,04403

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR