D-(+)-Biotin

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ALX460002G001 ALX460002G005
ENZOALX460002G001EA 50.6 EUR
ENZOALX460002G001 ENZOALX460002G005
D-(+)-Biotin
D-(+)-Biotin
Formule: C₁₀H₁₆N₂O₃S
Smeltpunt: 232…233 °C
Dichtheid: 1,41 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005541
CAS nummer: 58-85-5
EINECS: 200-399-3
Merck Index: 13,01231

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR