Human recombinant TNF alpha (soluble) (from E. coli)
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ALX522008C050
ENZOALX522008C050EA 479 EUR
ENZOALX522008C050
Human recombinant TNF alpha (soluble) (from E. coli)
Eiwitten en peptiden
Produced in E. coli. The extracellular domain of human TNF-α (tumor necrosis factor-α) (aa 85-223) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.