6-Cyano-7-nitroquinoxaline-2,3-dione ≥98% (by TLC)

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ALX550042M005 ALX550042M010 ALX550042M025 ALX550042M050
ENZOALX550042M005EA 85.8 EUR
ENZOALX550042M005 ENZOALX550042M010 ENZOALX550042M025 ENZOALX550042M050
6-Cyano-7-nitroquinoxaline-2,3-dione ≥98% (by TLC)
6-Cyano-7-nitroquinoxaline-2,3-dione

AMPA/kainate receptor antagonist


  • Formulation: Off-white to yellow powder.


Potent, competitive kainate/quisqualate (non-NMDA) receptor antagonist. For the more soluble form see CNQX . disodium salt (Prod. No. ALX-550-283).

Formule: C₉H₄N₄O₄
MDL Nummer: MFCD00069232
CAS nummer: 115066-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR