Serotonin hydrochloride

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ALX550328G001 ALX550328M050
ENZOALX550328G001EA 317 EUR
ENZOALX550328G001 ENZOALX550328M050
Serotonin hydrochloride
Serotonin hydrochloride
Formule: C₁₀H₁₃ClN₂O
MDL Nummer: MFCD00012686
CAS nummer: 153-98-0
UN: 2811
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR