Lipopolysaccharides (from S. typhimurium) S-form, TLRgrade™ ready-to-use

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

ALX581011L001 ALX581011L002
ENZOALX581011L001EA 224 EUR
ENZOALX581011L001 ENZOALX581011L002
Lipopolysaccharides (from S. typhimurium) S-form, TLRgrade™ ready-to-use
Lipopolysaccharides

TLR4 activator.


Smooth (S)-form LPS, isolated and purified from Salmonella typhimurium by modification of the phenol water extraction and PCP method, converted to the uniform salt form.


Specific activator of Toll-like receptor (TLR) 4. Does not activate TLR2 or other TLRs as determined with splenocytes and macrophages from TLR4 deficient mice. Smooth (S)-form LPS are commonly the preferred choice for whole animal studies, whereas Rough (R)-form LPS are primarily used in cellular in vitro activation studies.

Order Now

Specification Test Results

Concentration 1 mg/ml
Formulation Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
Purity Absence of detectable protein or DNA contaminants with agonistic TLR activity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR