Aflatoxin G1 ≥98% (by TLC)

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

ALX630104M001 ALX630104M005
ENZOALX630104M001EA 160 EUR
ENZOALX630104M001 ENZOALX630104M005
Aflatoxin G1 ≥98% (by TLC)
Aflatoxin G1

Potent hepatotoxic and hepatocarcinogenic mycotoxin.


Soluble in DMSO (10 mg/ml), methylene chloride (5 mg/ml) or methanol.

Formule: C₁₇H₁₂O₇
Bewaar temperatuur: Koelkast
CAS nummer: 1165-39-5
EINECS: 214-615-9
UN: 2811
ADR: 6.1,I
Merck Index: 14,00181

Order Now

Specification Test Results

Source Isolated from Aspergillus flavus
Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR