Native Hsv-2 antigen

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ALX6301270200
ENZOALX6301270200EA 678 EUR
ENZOALX6301270200
Native Hsv-2 antigen
Eiwitten en peptiden

From herpes simplex virus type 2, strain G. Inactivated by 0,1% Triton X-100.

New Product

Order Now

Specificaties

Protein/peptide name Hsv-2 Antigen
Environmentally Preferable
Protein/peptide type Inheems
Species
Bron
Zuiverheid ~80%
Formulering Liquid, PBS containing 0,1% Triton X-100

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR