Anti-STAT3 Mouse Monoclonal Antibody
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ALX804180C100
ENZOALX804180C100EA 500 EUR
ENZOALX804180C100
Anti-STAT3 Mouse Monoclonal Antibody
Antilichamen
The STAT proteins serve as both cytoplasmic signal transducers and nuclear activators of transcription. STAT3 is activated by tyrosine phosphorylation at residue Y705 in cells treated with interleukin-6 or EGF. Activated STAT3 can bind to DNA either as homodimer or as heterodimer with STAT1.
  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes the -AAPpYLKT-motif of activated human, mouse and dog STAT3 phosphorylated at Tyr705. Does not cross-react with the non-phosphorylated form of STAT3More Product Information