Anti-BIMS/EL/L Rat Monoclonal Antibody

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ALX804528C100
ENZOALX804528C100EA 533 EUR
ENZOALX804528C100
Anti-BIMS/EL/L Rat Monoclonal Antibody
Antilichamen
Host: Rat, Isotype: IgG2a

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes an epitope within aa 20-40 of human, mouse, rat, monkey and dog BimL, BimEL, and BimS. Also recognizes the six novel isoforms of human Bim, generated by alternative splicing and lacking the C-terminal hydrophobic region recently identified.

Type: Primary
Antigen: BIMS/EL/L
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR