Phosphothreonine detection set
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ALX850024KI01
ENZOALX850024KI01EA 569 EUR
ENZOALX850024KI01
Phosphothreonine detection set
Koppelings- en labelingssystemen voor eiwitten
This kit offers a set of three different monoclonal antibodies specific for threonine phosphorylation sites. Phosphorylation patterns in a given cell extract may differ when probed with different antibodies due to sequence specificity. Recognition therefore depends on two criteria: Phosphorylation and surrounding aa motif. If one of the two criteria is not fulfilled, the antibody will not detect the phosphorylation site.
Immunoprecipitation; Western Blot (20µl/lane (mini gel) for HRPO/DAB detection, or 5µl/lane (mini gel) for HRPO/ECL detection). Do not use milk/casein for blocking and dilution! Replace by BSA! We recommend to use 1% BSA, 1% PVP-10 (polyvinylpyrrolidone), 1% PEG 3500, 0.2% Tween 20 in 2x PBS.Optimal conditions must be determined individually for each application.
Bestelinformatie: Positive control included. Contents: Contains 25µg each of clones:1E11 (Prod. No. ALX-804-168)4D11 (Prod. No. ALX-804-169)14B3 (Prod. No. ALX-804-170)
Order Now
Specificaties
Environmentally Preferable
Host
Learn more