Caspase fluorometric substrate set

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ALX850229KI01
ENZOALX850229KI01EA 953 EUR
ENZOALX850229KI01
Caspase fluorometric substrate set
Eiwitten en peptiden

Fluorometric substrates for assaying activities of members of caspase family proteases. All substrates are provided in liquid ready-to-use form.


Bestelinformatie: Contents: Contains 125 µl (1mM) each of the following substrates:
- Caspase-1 Substrate, Ac-YVAD-AFC
- Caspase-2 Substrate, Ac-VDVAD-AFC
- Caspase-3 Substrate, Ac-DEVD-AFC
- Caspase-5 Substrate, Ac-WEHD-AFC
- Caspase-6 Substrate, Ac-VEID-AFC
- Caspase-8 Substrate, Ac-IETD-AFC
- Caspase-9 Substrate, Ac-LEHD-AFC

Order Now

Specificaties

Protein/peptide name
Environmentally Preferable
Species
Bron
Formulering

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR