RO 48-8071 ≥95% (by HPLC)

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BMLEI3420010 BMLEI3420050
ENZOBMLEI3420010EA 89.1 EUR
ENZOBMLEI3420010 ENZOBMLEI3420050
RO 48-8071 ≥95% (by HPLC)
RO 48-8071

Inhibits cholesterol biosynthesis at the level of 2,3-oxidosqualene cyclase (OSC). Blocks OSC and cholesterol synthesis in HepG2 cells in the nanomolar range. A novel lipid lowering agent.


  • Oxido-squalene cyclase inhibitor


Pale yellow solid.


Soluble in water (25 mg/ml)

Formule: C₂₃H₂₇BrFNO₂.C₄H₄O₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD05865242
CAS nummer: 189197-69-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5cb77a4ad0172ef14421d20dbff445f65403837&vid=8feafbbb2a008a3b4c0c79664ccc375959114654&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR