Crosstide
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BMLP1490001
ENZOBMLP1490001EA 165 EUR
ENZOBMLP1490001
Crosstide
Eiwitten en peptiden
Akt substrate
This peptide sequence, GRPRTSSFAEG, corresponds to the sequence of GSK3 surrounding the serine phosphorylated by MAP kinase activated protein kinase (MAPKAP kinase-1) and p70 ribosomal S6 kinase. Acts as a substrate for Akt (PKB), Km=4 µM. Useful in phosphocellulose kinase assays.