AIP
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BMLP2110001
ENZOBMLP2110001EA 138 EUR
ENZOBMLP2110001
AIP
Eiwitten en peptiden
CaM kinase inhibitor
AIP (autocamtide-2-related inhibitory peptide) is a highly specific and potent inhibitor of CaM kinase II. This peptide is derived from the CaM kinase II substrate autocamtide-2 (BML-P101) but alanine is substituted for threonine at the 9-position. AIP at 1 µM completely inhibited CaM kinase II but did not affect PKC, PKA, CaMKIV and other brain extract kinases. A cell-permeable, myristoylated version is available (BML-P212).