Nafamostat mesylate ≥98%

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BMLPI1430010 BMLPI1430050
ENZOBMLPI1430010EA 89.1 EUR
ENZOBMLPI1430010 ENZOBMLPI1430050
Nafamostat mesylate ≥98%
Nafamostat mesylate

Tryptase inhibitor


Competitive, reversible broad-spectrum serine proteinase inhibitor that can be a used as a specific tryptase inhibitor at very low concentrations. Has been used in vitro and in vivo.

Formule: C₁₉H₁₇N₅O₂
MDL Nummer: MFCD00941430
CAS nummer: 82956-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74636c826c7833c19afeaaf3c891280366abe4fc&vid=b39da665ee85f52496ebc349d89f965adec8dc76&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR