Human recombinant tyrosinase (from E. coli)
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BMLSE5350100
ENZOBMLSE5350100EA 396 EUR
ENZOBMLSE5350100
Human recombinant tyrosinase (from E. coli)
Enzymen
Produced in E. coli. Tyrosinase is a copper-containing enzyme that catalyzes four steps in the melanin and catecholamine synthesis pathways.
  • Oxidizes monophenols, but like catecholoxidase (EC 1.10.3.1), it can also oxidize o-diphenols to o-quinones
  • Involved in cancer, Parkinson’s, pigmentation diseases, Alzheimer’s and obesity
  • Much of the research and inhibitor screening has used mushroom tyrosinase
Like tyrosine hydroxylase (EC 1.13.16.2), it can catalyze the formation of L-DOPA in the brain and nervous system. However, mushroom tyrosinase interacts very differently with substrates and inhibitors compared to the human enzyme.
In addition, commercially available mushroom tyrosinase is not always of good purity. Recombinant versions of human tyrosinase amino acids 1-345 or 1-378, also available commercially, are not functional due the lack of critical catalytic and structural residues.