D-erythro-Dihydrosphingosine ≥98%

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BMLSL1250010 BMLSL1250050
ENZOBMLSL1250010EA 84.7 EUR
ENZOBMLSL1250010 ENZOBMLSL1250050
D-erythro-Dihydrosphingosine ≥98%
D-erythro-Dihydrosphingosine

Sphingosine precursor


Biosynthetic precursor of sphingosine. Inhibitor of protein kinase C (PKC). Blocks phospholipases A2 (PLA2) and the D-sphingosine precursor.

Formule: C₁₈H₃₉NO₂
MDL Nummer: MFCD07371371
CAS nummer: 764-22-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d59a66d01008d6ca93b2f13390c1279a23d0e904&vid=62e3a258b169096f69670c63b89d25becbab89c1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR