NGAL (mouse) ELISA kit

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BPDKIT042
ENZOBPDKIT042EA 964 EUR
ENZOBPDKIT042
NGAL (mouse) ELISA kit
Testen ELISAs

For the quantitative determination of NGAL in mouse urine, plasma, serum, tissue extracts, or culture supernatants.


  • Ready to use pre-coated plate and individual calibrators save time and reduce errors
  • Fast results from up to 40 samples in duplicate in just 4 hours per 96 well plate
  • Absorbance is read at 450 nm


A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in mouse samples. Absorbance is read at 450nm.


Leveringsinformatie: The kit contains a microwell plate coated with anti-NGAL (mouse) antibody (96 wells), 5X sample diluent, 2 containers of lyophilised mouse NGAL calibrators, 25X wash solution concentrate, biotinylated mouse NGAL antibody, HRP-streptavidin, TMB-substrate and stop solution.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR