Methylated DNA IP kit, BIOARRAY™
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ENZ450120010
ENZOENZ450120010EA 880 EUR
ENZOENZ450120010
Methylated DNA IP kit, BIOARRAY™
Testen Nucleïnezuurbepalingen
BIOARRAY™ Methylated DNA IP Kit features IP technology for the enrichment of 5-methylcytosine-containing DNA from any pool of fragmented genomic DNA for use in genome-wide methylation analysis in the completion of diverse downstream applications.
  • Highly specific antibody is included to distinguish methylated DNA from pool
  • DNA enrichment for large-scale DNA methylation analysis
  • Easy to use with reproducible results
The kit includes a highly specific anti-5-methylcytosine monoclonal antibody for the capture and separation of methylated DNA from non-methylated DNA in only a few hours.
Leveringsinformatie: Kit contains Protein A Beads, Anti-5 methylcytosine, Control DNA, Control primer 1 , Control primer 2, IP Buffer, DNA denaturing Buffer, Control mouse DNA (methylated/nonmethylated), DNA elution buffer, magnetic rods.
Order Now
Specificaties
Environmentally Preferable