MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) 98 HPLC_ASSAY_METHOD, ultrapure

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (6-Methoxyquinolinio)acetic acid ethyl ester bromide, 1-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide, 1-(2-Ethoxy-2-oxoethyl)-6-methoxyquinolin-1-ium bromide, MQAE

ENZ52156
ENZOENZ52156EA 165 EUR
ENZOENZ52156
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) 98 HPLC_ASSAY_METHOD, ultrapure
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide)

Chloride dye


MQAE is a fluorescent chloride ion indicator that is more sensitive and selective than 36Cl and microelectrode-based methods for chloride measurement in cells. MQAE is a fluorescent indicator that is quenched via collision with chloride. The probe is suitable for detection of chloride concentrations in liposomes, reconstituted membranes or in extracellular medium. As a UV-excitable indicator, MEQ can be detected using an argon-ion laser for confocal microscopy and flow cytometry applications. MQAE has greater sensitivity than SPQ and higher fluorescence quantum yield. The ester group of MQAE may slowly hydrolyze inside cells, resulting in a change in its fluorescence response. Wavelength Maxima: Excitation 350 nm, Emission 460 nm.

Formule: C₁₄H₁₆NO₃Br
MW: 326.19 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00467854
CAS nummer: 162558-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddbfcf8916487f04195c827f2b4148066cb2fd10&vid=022ec38afe0064e741ce593c65e93b0e3e369a7a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR