27-Hydroxycholesterol ≥98% (by HPLC)

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ENZCHM1020001 ENZCHM1020005
ENZOENZCHM1020001EA 72.6 EUR
ENZOENZCHM1020001 ENZOENZCHM1020005
27-Hydroxycholesterol ≥98% (by HPLC)
27-Hydroxycholesterol

Cholesterol metabolite linked to breast cancer, neurodegenerative disease and metabolism.


27-Hydroxycholesterol (27-HC) is a metabolite resulting from the action of sterol 27-Hydroxylase on cholesterol. It is an endogenous selective estrogen receptor modulator and ligand for the liver X receptor (LXR). 27-Hydroxycholesterol plays a role in Alzheimer's disease, oxidative stress, metabolism, cell-signaling, and heart disease.


Soluble in DMSO (up to 10 mg/ml with warming) or in ethanol (up to 10 mg/ml). Insoluble in water.

Formule: C₂₇H₄₆O₂
Bewaar temperatuur: Vriezer
CAS nummer: 20380-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41246d5c0320d9bd08b6caa11cb7b09255159684&vid=bcab6e77cf6f634830fce6305eaad339d433c0de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR