ApoE ELISA kit, grow cells
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ENZKIT1340001
ENZOENZKIT1340001EA 901 EUR
ENZOENZKIT1340001
ApoE ELISA kit, grow cells
Testen ELISAs
Highly sensitive ApoE ELISA kit enabling detection of ApoE in serum, plasma, cell culture supernates, and tissue homogenates in 3,5 hours.
  • Sensitive measurement of ApoE, detecting as little as 14,063 pg/ml
  • Negligible reactivity to similar proteins.
  • High throughput with results in 3,5 hours.
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of ApoE in serum, plasma, cell culture supernates and tissue homogenates in human samples.
The ApoE ELISA kit is a colorimetric immunometric immunoassay kit with results in 3,5 hrs.
Bestelinformatie: Contains Microtiter Plate, Standard, Sample Buffer, Wash Buffer Concentrate, Antibody, Antibody Buffer, Conjugate, Conjugate Buffer Substrate, and Stop Solution.