epT.I.P.S.® Reloads, Pipette Tips
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0030073.819 0030073.371 0030073.398 0030073.410 0030073.436 0030073.452 0030073.479 0030073.495 0030073.517 0030073.630 0030073.649 0030073.754 0030073.770 0030073.797 0030073.835 0030073.851 0030073.878 0030073.894
EPPE0030073.819EA 81.8 EUR
EPPE0030073.819 613-6068 613-6069 613-6070 613-6071 613-6072 613-6073 613-6074 613-6075 613-6076 EPPE0030073.649 EPPE0030073.754 EPPE0030073.770 EPPE0030073.797 EPPE0030073.835 EPPE0030073.851 EPPE0030073.878 EPPE0030073.894
epT.I.P.S.® Reloads, Pipette Tips
Pipettips
Eppendorf epT.I.P.S.® Reloads offer high-precision pipette tips in 'Eppendorf Quality' and 'PCR clean' purity grades. Eco-friendly, cost-effective, and designed for seamless compatibility with Eppendorf Box 2.0, they ensure accuracy and sustainability in labs.
  • Reduced waste when compared with disposable racks
  • Refill trays can be autoclaved up to 121 °C, in packaging and without foil, for subsequent use
  • Depending on tip size, the refill system is packaged as either dual-sided or in stack form
  • Available in purity grades of Eppendorf Quality and PCR clean
More Product Information