epT.I.P.S.® Sets, Pipette Tips
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0030076.290 0030076.303 0030076.311 0030076.320 0030076.338 0030076.346 0030076.354 0030076.362
EPPE0030076.290EA 70.2 EUR
EPPE0030076.290 EPPE0030076.303 EPPE0030076.311 EPPE0030076.320 EPPE0030076.338 EPPE0030076.346 EPPE0030076.354 EPPE0030076.362
epT.I.P.S.® Sets, Pipette Tips
Pipettips
Eppendorf epT.I.P.S.® Sets, in 'Eppendorf Quality' grade, combine the convenience of a reusable Box 2.0 with high-quality reloadable pipette tips, offering a sustainable, efficient solution for precise and organised pipetting in laboratories.
  • Contamination-free transfer of trays to stable working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and pipette
  • Tips can be attached to pipette from refill trays
  • Reloads and Boxes can be autoclaved at 121 °C for subsequent use
  • epT.I.P.S.® Box 2.0 and epT.I.
More Product Information