Bottles, narrow neck, round, HDPE, with thread
Leverancier: EP SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
013-1000BP 013-0250BP 013-0500BP
EPSC013-1000BPEA 206 EUR
EPSC013-1000BP EPSC013-0250BP EPSC013-0500BP
Bottles, narrow neck, round, HDPE, with thread
Flessen Kunststof flessen
HDPE, translucent.
  • Narrow neck for precision pouring
  • Slim bottle design for easy grip and handling
  • For general purpose sampling where cleaning and certification is not required or ready for own cleaning and certification procedures
Narrow neck round bottle are general purpose containers which are unprocessed and uncertified.
Bestelinformatie: Bulk packed, caps must be ordered separately.