Water sampling bottles, glass, narrow neck, Boston type, round
Leverancier: EP SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
329-0500 329-1000 349-0500 329-0250 349-0125 229-0125 229-0250 229-0500 229-1000
EPSC329-0500EA 104 EUR
EPSC329-0500 EPSC329-1000 EPSC349-0500 EPSC329-0250 EPSC349-0125 EPSC229-0125 EPSC229-0250 EPSC229-0500 EPSC229-1000
Water sampling bottles, glass, narrow neck, Boston type, round
Flessen Watermonsternameflessen
These bottles, made of Type III soda-lime glas, clear or amber, with cap, are recommended for collecting water samples for variety of organic and inorganic target analytes where light protection is not required.
  • With PTFE-lined white PP closed top cap (assembled on bottles)
  • Certified to meet EPA performance based specifications for organics, pesticides and metal anlyses or unprocessed and readily assembled for own cleaning procedure
  • Each jar individually barcoded with lot number and unique container number
Leveringsinformatie: Supplied withMore Product Information
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage
Milieu certificering NAR Certificate of Retirement