Sample vials, with cap
Leverancier: EP SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
140-60C S346-0040 141-40A 340-40C 339-20C 340-40C/DB 340-60C 339-20A S346-0060 S336-0060 S136-0060 S236-0060 C346-0020
EPSC140-60CEA 878 EUR
EPSC140-60C EPSCS346-0040 EPSC141-40A EPSC340-40C EPSC339-20C EPSC340-40C/DB EPSC340-60C EPSC339-20A EPSCS346-0060 EPSCS336-0060 EPSCS136-0060 EPSCS236-0060 EPSCC346-0020
Sample vials, with cap
flesjes
Clear or amber sample vials made from borosilicate glass.
  • Designed for water sample collection for volatile organic analysis (VOA)
  • Optimised for use with automation
  • With open-top white PP cap bonded with 3,17 mm thin PTFE-lined silicone septum
  • Septum permanently bonded to the cap for secure sealing
  • Vial shoulder slope optimized to prevent air bubble entrapment
Processed or certified to meet or exceed EPA Performance Based Specifications for volatile organic analysis with certificate. Processed vials are processed in the same manner as certified vialsMore Product Information