Caps, closed top
Leverancier: EP SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
B167-2800BL B167-3300BL B167-3330BL B167-3800BL B167-5300BL B167-3830BL B167-5800BL B167-7000BL B167-8300BL B167-8900BL B167-4500BL
EPSCB167-2800BLEA 57.9 EUR
EPSCB167-2800BL EPSCB167-3300BL EPSCB167-3330BL EPSCB167-3800BL EPSCB167-5300BL EPSCB167-3830BL EPSCB167-5800BL EPSCB167-7000BL EPSCB167-8300BL EPSCB167-8900BL EPSCB167-4500BL
Caps, closed top
Sluitingen Flesdoppen
PP, white.
  • Very good chemical resistance
  • Very good temperature resistance
Unprocessed caps are ideal for environmental sample bottles, jars, jugs and vials.
Certificaten: Manufactured under an ISO certified quality management system