Signs, floor signs, DuraStripe ®, X-Treme, letters and numbers
Leverancier: ERGOMAT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4070058XL9R 4070058XL20G 4070058XN9G 4070058XN20R 4070058XN9Y 4070058XN20Y
ERGO4070058XL9REA 93 EUR
ERGO4070058XL9R ERGO4070058XL20G ERGO4070058XN9G ERGO4070058XN20R ERGO4070058XN9Y ERGO4070058XN20Y
Signs, floor signs, DuraStripe ®, X-Treme, letters and numbers
Pictogrammen
DuraStripe® is the easiest, fastest and least expensive floor marking system for industrial floors. The durability and vast size of the DuraStripe program makes it the only total floor marking solution available on the market today. Economic than painting the floors.
  • Ideal for less than perfect floors
  • Simple and easy peel-&-stick installation
  • No dry time, cure time, fumes or messy clean up to limit productivity