Cleanroom anti-fatigue mats, Ergomat® Nitril and Nitril smooth
Leverancier: ERGOMAT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ENS0420 ENS0412 ENS0406 ENS0320 ENS0220 ENS0209 ENS0206 ENS0203 EN0203 EN0420 EN0412 EN0406 EN0320 EN0306 EN0304 EN0220 EN0206 EN0209 ENS0404 ENS0409 EN0303 EN0212 EN0309 EN0312 EN0204 EN0404 EN0409 ENS0303 ENS0212 ENS0309 ENS0312 ENS0306
115-0894EA 2590 EUR
115-0894 115-0893 115-0892 115-0891 115-0888 115-0887 115-0886 115-0885 115-0865 115-0874 115-0873 115-0872 115-0871 115-0870 115-0869 115-0868 115-0866 115-0867 ERGOENS0404 ERGOENS0409 ERGOEN0303 ERGOEN0212 ERGOEN0309 ERGOEN0312 115-2247 ERGOEN0404 ERGOEN0409 ERGOENS0303 ERGOENS0212 ERGOENS0309 ERGOENS0312 115-2248
Cleanroom anti-fatigue mats, Ergomat® Nitril and Nitril smooth
Vloermatten Anti-vermoeidheidsmatten
The Ergomat® Nitril and Nitril Smooth are made from pure nitrile rubber and is designed to increase comfort while reducing trip hazards, body pain, quality issues and sick leave. It is ideal for areas where heat, oil and petrochemical resistance is crucial.
  • Choice of smooth or coned upper surface
  • Can be autoclaved (610×915 mm only) and features great antibacterial cleaning properties
  • Chemical exposure will not affect the state of the mat; no curling up, swelling, or expanding
  • Moulded, bevelled edges prevent curling, eliminating trip hazards
  • Raw, virgin
More Product Information
Order Now
Specificaties
Environmentally Preferable