PCR workstation, CaptairBio
Leverancier: ERLAB CAPTAIRE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AE206Z00011C21
ERLAAE206Z00011C21EA 3779 EUR
ERLAAE206Z00011C21
PCR workstation, CaptairBio
Zuurkasten en behuizingen PCR-werkstations
Designed to perform non-pathogenic handlings, these enclosures has been created for handlings involving a high sample cross contamination risk such as sample preparations before thermocycling, Post PCR DNA sequencing revelation / separation, in vitro fertilization, cellular cultures, vegetal biology, preparation of sterile solutions.
Please complete the quote request to enable us to assist you in finding the right unit for your application.
https://nl.cmd2.vwr.com/bin/public/fc/main?f=Y5&pimid=26080247
  • The worksurface made of stainless steel makes the cleaning easy
  • Total openings for