Conductivity electrode, Eutech
Leverancier: EUTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
93X546101
EUTE93X546101EA 328 EUR
EUTE93X546101
Conductivity electrode, Eutech
Elektroden Geleidbaarheidselektroden
Conductivity/temperature sensor with epoxy body and stainless steel rings. For use with the following Eutech meters: CON 700 / PC 700 / CON 2700 / PC 2700.