Anti-SLC47A1 Goat Polyclonal Antibody

Leverancier: Everest Biotech
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

EB09191
EVBIEB09191EA 335 EUR
EVBIEB09191
Anti-SLC47A1 Goat Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: SLC47A1 (solute carrier family 47 (multidrug and toxin extrusion), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Specificaties

antigeen symbool SLC47A1
Environmentally Preferable
Type antilichaam Primair,solute carrier family 47 (multidrug and toxin extrusion), member 1
Type antilichaam Primair,solute carrier family 47 (multidrug and toxin extrusion), member 1
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Goat
Isotype IgG
Western blot Yes
ELISA Yes
Epitoop internal
Kruisadsorptie none
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR