Hand cleansers, Estesol® premium

Leverancier: Deb-STOKO
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

22350
EVON22350EA 187.2 EUR
EVON22350
Hand cleansers, Estesol® premium
Huidverzorging Zepen en huidreinigingsmiddelen
Liquid skin cleanser for light to medium-duty contamination. Ideal for highly-stressed and sensitive skin.

  • Contains skin compatible sugar surfactants and natural moisturisers
  • Skin friendly pH
  • Subtly scented

Colour: Clear

Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR