Avantor® ACE® Excel® C18-PFP, HPLC/UHPLC Columns, Analytical, 5 µm
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
EXL-1210-1002UEA 826 EUR
EXL-1210-1002U EXL-1210-1003U EXL-1210-1046U EXL-1210-1502U EXL-1210-1503U EXL-1210-1546U EXL-1210-2502U EXL-1210-2503U EXL-1210-2546U EXL-1210-0502U EXL-1210-0503U EXL-1210-0546U
Avantor® ACE® Excel® C18-PFP, HPLC/UHPLC Columns, Analytical, 5 µm
Kolommen voor chromatografie
Avantor® ACE® Excel® C18-PFP phase utilizes a specially developed ligand combining a C18 chain with integral PFP functionality. This results in a phase that maintains the hydrophobic, stability and low bleed characteristics of leading C18 phases, yet provides the multiple retention mechanisms of a PFP phase.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1,7 to 5 µm) and complementary phases
  • Also useful for analytes with π-bonding or
More Product Information