Exsil ODS, HPLC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
EXODS1-5-250AEA 527 EUR
EXODS1-5-250A EXODS-10-250P EXODS-5-100AM EXODS-5-10CP3 EXODS-5-250AF EXODS-5-250P
Exsil ODS, HPLC Columns
Kolommen voor chromatografie
Exsil HPLC columns are a useful alternative to the classical spherical HPLC silicas.
  • Spherical porous silica
  • Competitively priced
  • Hichrom high efficiency
Exsil 100, the standard material, is supplied in a range of particle sizes and bonded phases to cover a wide range of applications.