Aqueous mounting medium, Fluoromount™, Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F4680-25ML
F4680-25MLEA 139 EUR
F4680-25ML
Aqueous mounting medium, Fluoromount™, Sigma-Aldrich®
Kleurstoffen Microscopy and staining reagents
Fluoromount™ Aqueous Mounting Medium for use with fluorescent dye-stained tissue.
Used for mounting:
- Coverslips on slides for immunofluorescence microscopic examination of PC12 and Hela cells
- Hippocampus neurons cells for confocal microscope studies
- Brain sections for immunofluorescence staining
Order Now
Specificaties
Environmentally Preferable