Avantor® Genesis, HPLC Columns, C18
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FK15963EEA 0 EUR
FK15963E FL5963E FM5963E FM15963E FK5960E FK10960E FL5960E FM5960E FM10960E FM15960E FM25960E 8P25960 FH15960E FH1960-2/N FH1963-2/N GEN-5C18-125D GEN-4C18-125D GEN-4C18-125DT GEN-4C18-150DE FH25960E FL25960E HICHGEN-4C18-100DE HICHGEN-5C18-150D HICHGEN-4C18-300DE GEN-5C18-250D
Avantor® Genesis, HPLC Columns, C18
Kolommen voor chromatografie
Avantor® Genesis phases are based on high purity metal-free spherical silica.
  • Good peak shape and reproducibility
  • Long column lifetime
They are suitable for the analysis of a wide range of analytes.
Stainless steel columns.