Colour Lab Tapes
Leverancier: GA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PAT-13WH PAT-13YE PAT-13LBL PAT-13RE PAT-13GA PAT-13PI PAT-13GR PAT-13OR PAT-13PU PAT-13GS PAT-18WH PAT-18YE PAT-18LBL PAT-18RE PAT-18GA PAT-18PI PAT-18GR PAT-18OR PAT-18PU PAT-18GS PAT-24WH PAT-24YE PAT-24LBL PAT-24RE PAT-24GA PAT-24PI PAT-24GR PAT-24OR PAT-24PU PAT-24GS
GAINPAT-13WHEA 18.2 EUR
GAINPAT-13WH GAINPAT-13YE GAINPAT-13LBL GAINPAT-13RE GAINPAT-13GA GAINPAT-13PI GAINPAT-13GR GAINPAT-13OR GAINPAT-13PU GAINPAT-13GS GAINPAT-18WH GAINPAT-18YE GAINPAT-18LBL GAINPAT-18RE GAINPAT-18GA GAINPAT-18PI GAINPAT-18GR GAINPAT-18OR GAINPAT-18PU GAINPAT-18GS GAINPAT-24WH GAINPAT-24YE GAINPAT-24LBL GAINPAT-24RE GAINPAT-24GA GAINPAT-24PI GAINPAT-24GR GAINPAT-24OR GAINPAT-24PU GAINPAT-24GS
Colour Lab Tapes
Tape
Colour laboratory tape is available in a wide range of colours and widths, for easy identification and colour-coding.
  • Temperature range: –80 to +121 °C
  • Ideal for all general-use applications in the lab
  • Continuous tape format allows use of as much or little as necessary
  • Resistant to gamma irradiation (tested to up to 50 kGy)
  • Available in a wide range of colors and widths, for easy identification and colour-coding
Available in an assortment of eye-catching colours and various widths. Lab tape is perfect for colour-coding and general lab use. Can beMore Product Information
Order Now
Specificaties
Environmentally Preferable